ENA413172440 Enamine

N1-(pyrazin-2-yl)ethane-1,2-diamine

  • MDL number MFCD09041693
  • Cas Number : 440102-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33535
Availability details for
Quantity
  • Availability
  • Add to Cart