ENA413172461 Enamine

5-(azepane-1-sulfonyl)-2-chloroaniline

  • MDL number MFCD02704398
  • Cas Number : 326916-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-33956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33956
Availability details for
Quantity
  • Availability
  • Add to Cart