ENA413172478 Enamine

1-N-(4-methoxyphenyl)benzene-1,2-diamine

  • MDL number MFCD03478356
  • Cas Number : 25914-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34037

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34037
Availability details for
Quantity
  • Availability
  • Add to Cart