ENA413172480 Enamine

(3R)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD00144038
  • Cas Number : 103733-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300...302
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34053

Safety Information

safety_statement 26-36
msds https://www.enaminestore.com/catalog/EN300-34053
Availability details for
Quantity
  • Availability
  • Add to Cart