ENA413172484 Enamine

5-chloro-2-(morpholin-4-yl)aniline

  • MDL number MFCD02934447
  • Cas Number : 90875-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34073

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-34073
Availability details for
Quantity
  • Availability
  • Add to Cart