ENA413172503 Enamine

2-[(ethylamino)methyl]-6,7-dimethoxy-3,4-dihydroquinazolin-4-one

  • MDL number MFCD04623501
  • Cas Number : 730976-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-206
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34132
Availability details for
Quantity
  • Availability
  • Add to Cart