ENA413172505 Enamine

2-(propylamino)-N-(2,3,4-trifluorophenyl)acetamide

  • MDL number MFCD06372774
  • Cas Number : 796106-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34135
Availability details for
Quantity
  • Availability
  • Add to Cart