ENA413172506 Enamine

2-(methylamino)-N-propylacetamide

  • MDL number MFCD05270871
  • Cas Number : 901273-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34139
Availability details for
Quantity
  • Availability
  • Add to Cart