ENA413172509 Enamine

(2,3-dihydro-1,4-benzodioxin-2-ylmethyl)(ethyl)amine

  • MDL number MFCD01659511
  • Cas Number : 21398-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34143

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34143
Availability details for
Quantity
  • Availability
  • Add to Cart