ENA413172520 Enamine

2-phenyl-2-(2-phenyl-1H-indol-3-yl)ethan-1-amine

  • MDL number MFCD03988751
  • Cas Number : 102441-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34174
Availability details for
Quantity
  • Availability
  • Add to Cart