ENA413172525 Enamine

7-bromo-2,3-dihydro-1,4-benzodioxin-6-amine

  • MDL number MFCD00462568
  • Cas Number : 368839-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34186
Availability details for
Quantity
  • Availability
  • Add to Cart