ENA413172531 Enamine

[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl](methyl)amine

  • MDL number MFCD01659517
  • Cas Number : 2242-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34200
Availability details for
Quantity
  • Availability
  • Add to Cart