ENA413172535 Enamine

2-[(ethylamino)methyl]-3,4-dihydroquinazolin-4-one

  • MDL number MFCD04621553
  • Cas Number : 143993-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34208
Availability details for
Quantity
  • Availability
  • Add to Cart