ENA413172539 Enamine

2-{[(furan-2-yl)methyl]amino}-N-[2-(trifluoromethyl)phenyl]acetamide

  • MDL number MFCD04206454
  • Cas Number : 838093-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-183
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34218
Availability details for
Quantity
  • Availability
  • Add to Cart