ENA413172546 Enamine

4-[1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-1,3-thiazol-2-amine

  • MDL number MFCD03474978
  • Cas Number : 379240-65-0
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-1,3-thiazol-2-amine
Inchi Code 1S/C15H14FN3S/c1-9-7-13(14-8-20-15(17)18-14)10(2)19(9)12-5-3-11(16)4-6-12/h3-8H,1-2H3,(H2,17,18)
Inchi Key FWULYMDBWAYWSX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-34239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34239
Availability details for
Quantity
  • Availability
  • Add to Cart