ENA413172559 Enamine

7-chloro-2-[(propylamino)methyl]-3,4-dihydroquinazolin-4-one

  • MDL number MFCD06347927
  • Cas Number : 900640-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34266
Availability details for
Quantity
  • Availability
  • Add to Cart