ENA413172560 Enamine

2-[4-(trifluoromethyl)phenyl]ethan-1-amine

  • MDL number MFCD01529875
  • Cas Number : 775-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34269

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34269
Availability details for
Quantity
  • Availability
  • Add to Cart