ENA413172578 Enamine

[(3,4-dimethoxyphenyl)methyl](ethyl)amine

  • MDL number MFCD04491618
  • Cas Number : 40171-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34324
Availability details for
Quantity
  • Availability
  • Add to Cart