ENA413172583 Enamine

1-[(4-chlorophenyl)methyl]piperidin-4-amine

  • MDL number MFCD07365309
  • Cas Number : 78471-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34342
Availability details for
Quantity
  • Availability
  • Add to Cart