ENA413172593 Enamine

5,6-dimethyl-2-[(piperazin-1-yl)methyl]-1H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD13358931
  • Cas Number : 900640-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34408
Availability details for
Quantity
  • Availability
  • Add to Cart