ENA413172600 Enamine

methyl[1-(3-nitrophenyl)ethyl]amine

  • MDL number MFCD07366362
  • Cas Number : 118949-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34461

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34461
Availability details for
Quantity
  • Availability
  • Add to Cart