ENA413172601 Enamine

methyl 1-[(4-aminobenzene)sulfonyl]piperidine-3-carboxylate

  • MDL number MFCD07366424
  • Cas Number : 901273-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34464

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34464
Availability details for
Quantity
  • Availability
  • Add to Cart