ENA413172603 Enamine

[(1-benzyl-1H-pyrazol-4-yl)methyl](methyl)amine

  • MDL number MFCD07686045
  • Cas Number : 956438-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-207
Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34477
Availability details for
Quantity
  • Availability
  • Add to Cart