ENA413172604 Enamine

2-(piperidin-1-ylcarbonyl)-3,4-dihydro-2H-1,4-benzoxazine

  • MDL number MFCD07366374
  • Cas Number : 92292-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34479

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34479
Availability details for
Quantity
  • Availability
  • Add to Cart