ENA413172606 Enamine

2-(4-methylpiperazin-1-yl)-5-(piperidine-1-sulfonyl)aniline

  • MDL number MFCD04620406
  • Cas Number : 901273-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-34485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34485
Availability details for
Quantity
  • Availability
  • Add to Cart