ENA413172616 Enamine

N-[(4-chlorophenyl)methyl]-2-(ethylamino)acetamide

  • MDL number MFCD05263138
  • Cas Number : 743444-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34508
Availability details for
Quantity
  • Availability
  • Add to Cart