ENA413172624 Enamine

1-[(2-methyl-1,3-thiazol-4-yl)methyl]piperazine

  • MDL number MFCD05864526
  • Cas Number : 880361-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34570
Availability details for
Quantity
  • Availability
  • Add to Cart