ENA413172627 Enamine

1-(pyridine-4-carbonyl)piperazine

  • MDL number MFCD01829015
  • Cas Number : 39640-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-34607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34607
Availability details for
Quantity
  • Availability
  • Add to Cart