ENA413172631 Enamine

N-(thiophen-2-ylmethyl)-2,3-dihydro-1,4-benzodioxin-6-amine

  • MDL number MFCD04562809
  • Cas Number : 869950-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34614

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34614
Availability details for
Quantity
  • Availability
  • Add to Cart