ENA413172639 Enamine

5,8-dimethoxy-1,2,3,4-tetrahydroisoquinolin-4-ol

  • MDL number MFCD07366383
  • Cas Number : 102073-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199-200
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-34777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34777
Availability details for
Quantity
  • Availability
  • Add to Cart