ENA413172643 Enamine

2-(ethylamino)-N-(thiophen-2-ylmethyl)acetamide

  • MDL number MFCD05263175
  • Cas Number : 743444-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34796

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34796
Availability details for
Quantity
  • Availability
  • Add to Cart