ENA413172666 Enamine

[(oxolan-2-yl)methyl][(thiophen-2-yl)methyl]amine

  • MDL number MFCD02026614
  • Cas Number : 351436-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35099
Availability details for
Quantity
  • Availability
  • Add to Cart