ENA413172673 Enamine

3-(aminomethyl)-N-methylbenzamide

  • MDL number MFCD09048986
  • Cas Number : 515131-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35201
Availability details for
Quantity
  • Availability
  • Add to Cart