ENA413172683 Enamine

4-[2-amino-1-(1H-indol-3-yl)ethyl]-N,N-dimethylaniline

  • MDL number MFCD04621506
  • Cas Number : 730949-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35275
Availability details for
Quantity
  • Availability
  • Add to Cart