ENA413172686 Enamine

4-(1,3-thiazol-2-yl)aniline

  • MDL number MFCD06655097
  • Cas Number : 193017-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35288

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35288
Availability details for
Quantity
  • Availability
  • Add to Cart