ENA413172687 Enamine

1-[(2,4-dimethylbenzene)sulfonyl]piperazine

  • MDL number MFCD03444523
  • Cas Number : 524711-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35290
Availability details for
Quantity
  • Availability
  • Add to Cart