ENA413172719 Enamine

methyl 2-(3-aminophenoxy)acetate

  • MDL number MFCD09047922
  • Cas Number : 158196-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35991
Availability details for
Quantity
  • Availability
  • Add to Cart