ENA413172731 Enamine

2-methyl-2-(1,2,3,4-tetrahydroisoquinolin-2-yl)propan-1-amine

  • MDL number MFCD05201595
  • Cas Number : 891642-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37061

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37061
Availability details for
Quantity
  • Availability
  • Add to Cart