ENA413172751 Enamine

1-(5-phenyl-1,3,4-oxadiazol-2-yl)piperazine

  • MDL number MFCD08444673
  • Cas Number : 923107-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39363
Availability details for
Quantity
  • Availability
  • Add to Cart