ENA413172755 Enamine

1-(2-ethoxyphenyl)propan-1-amine

  • MDL number MFCD09735595
  • Cas Number : 473732-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39667
Availability details for
Quantity
  • Availability
  • Add to Cart