ENA413172761 Enamine

2-{methyl[(propan-2-ylcarbamoyl)methyl]amino}acetic acid

  • MDL number MFCD07366360
  • Cas Number : 900641-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39977

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39977
Availability details for
Quantity
  • Availability
  • Add to Cart