ENA413172762 Enamine

4-(2H-1,2,3-triazol-2-yl)aniline

  • MDL number MFCD20486489
  • Cas Number : 52708-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39982

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39982
Availability details for
Quantity
  • Availability
  • Add to Cart