ENA413172764 Enamine

4-[4-(aminomethyl)phenyl]phenol

  • MDL number MFCD06212573
  • Cas Number : 60277-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40029
Availability details for
Quantity
  • Availability
  • Add to Cart