ENA413172768 Enamine

1-(pyridine-4-carbonyl)-1,4-diazepane

  • MDL number MFCD09047178
  • Cas Number : 199938-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40204
Availability details for
Quantity
  • Availability
  • Add to Cart