ENA413172775 Enamine

N1-(4-methylphenyl)benzene-1,2-diamine

  • MDL number MFCD00616510
  • Cas Number : 91806-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40549
Availability details for
Quantity
  • Availability
  • Add to Cart