ENA413172831 Enamine

2-[(2-aminophenyl)amino]ethan-1-ol

  • MDL number MFCD06335078
  • Cas Number : 4926-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40657

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40657
Availability details for
Quantity
  • Availability
  • Add to Cart