ENA413172903 Enamine

6-amino-7-fluoro-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD12173926
  • Cas Number : 1155000-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41394
Availability details for
Quantity
  • Availability
  • Add to Cart