ENA413172914 Enamine

tert-butyl 1-amino-6-azaspiro[2.5]octane-6-carboxylate

  • MDL number MFCD19440981
  • Cas Number : 1233323-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41917

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41917
Availability details for
Quantity
  • Availability
  • Add to Cart