ENA413172923 Enamine

4-methylquinolin-1-ium-1-olate

  • MDL number MFCD01716318
  • Cas Number : 4053-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-116
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42294
Availability details for
Quantity
  • Availability
  • Add to Cart