ENA413172935 Enamine

1-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole

  • MDL number MFCD00046879

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42800
Availability details for
Quantity
  • Availability
  • Add to Cart